Product Details

SNP ID
rs114662992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52460894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATCCACTCCCTTTTCAGGCTCA[C/T]CTTGAAGTAACCTGTGGATTATGGT
Phenotype
MIM: 615507 MIM: 191040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NISCH PubMed Links

Gene Details

Gene
NISCH
Gene Name
nischarin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276293.1 Intron NP_001263222.1
NM_001276294.1 Intron NP_001263223.1
NM_007184.3 Intron NP_009115.2
XM_005264839.2 Intron XP_005264896.1
XM_006712955.2 Intron XP_006713018.1
XM_017005643.1 Intron XP_016861132.1
Gene
TNNC1
Gene Name
troponin C1, slow skeletal and cardiac type
There are no transcripts associated with this gene.

View Full Product Details