Product Details

SNP ID
rs114723180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9367448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCAGTTGTTAGGACAACAGCT[C/T]TGTCTCCAATATGTATTTAGCATCT
Phenotype
MIM: 606525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRGAP3 PubMed Links

Gene Details

Gene
SRGAP3
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
There are no transcripts associated with this gene.

Gene
THUMPD3
Gene Name
THUMP domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114092.1 Intron NP_001107564.1
NM_015453.2 Intron NP_056268.2
XM_005265021.4 Intron XP_005265078.1
XM_005265022.4 Intron XP_005265079.1
XM_005265023.3 Intron XP_005265080.1
XM_005265024.4 Intron XP_005265081.1
XM_006713086.3 Intron XP_006713149.1
XM_011533570.2 Intron XP_011531872.1
XM_011533571.2 Intron XP_011531873.1
XM_017006113.1 Intron XP_016861602.1
XM_017006114.1 Intron XP_016861603.1
XM_017006115.1 Intron XP_016861604.1
XM_017006116.1 Intron XP_016861605.1
XM_017006117.1 Intron XP_016861606.1
XM_017006118.1 Intron XP_016861607.1
XM_017006119.1 Intron XP_016861608.1

View Full Product Details