Product Details

SNP ID
rs114562652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73078606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTACATTATAGTTAAATGCATT[A/C]AGTTAATTTCTAGAGAGCAGGACAG
Phenotype
MIM: 615929 MIM: 610428
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD17 PubMed Links

Gene Details

Gene
ANKRD17
Gene Name
ankyrin repeat domain 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286771.2 Intron NP_001273700.1
NM_015574.1 Intron NP_056389.1
NM_032217.4 Intron NP_115593.3
NM_198889.2 Intron NP_942592.1
XM_005265671.3 Intron XP_005265728.1
XM_005265672.3 Intron XP_005265729.1
XM_005265673.3 Intron XP_005265730.1
XM_017008011.1 Intron XP_016863500.1
XM_017008012.1 Intron XP_016863501.1
XM_017008013.1 Intron XP_016863502.1
XM_017008014.1 Intron XP_016863503.1
XM_017008015.1 Intron XP_016863504.1
XM_017008016.1 Intron XP_016863505.1
XM_017008017.1 Intron XP_016863506.1
Gene
COX18
Gene Name
COX18, cytochrome c oxidase assembly factor
There are no transcripts associated with this gene.

View Full Product Details