Product Details
- SNP ID
-
rs111621148
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:176626512 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAGGCCTGTTCCTTGGTTTTTTC[A/G]GCCACTGGAGCAGGGAGGGGGTTGA
- Phenotype
-
MIM: 602569
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EIF4E1B
PubMed Links
Gene Details
- Gene
- EIF4E1B
- Gene Name
- eukaryotic translation initiation factor 4E family member 1B
There are no transcripts associated with this gene.
- Gene
- MIR4281
- Gene Name
- microRNA 4281
There are no transcripts associated with this gene.
- Gene
- SNCB
- Gene Name
- synuclein beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001502.2 |
459 |
Silent Mutation |
GCC,GCT |
A56A |
NP_001001502.1 |
NM_001318034.1 |
459 |
Silent Mutation |
GCC,GCT |
A42A |
NP_001304963.1 |
NM_001318035.1 |
459 |
Silent Mutation |
GCC,GCT |
A56A |
NP_001304964.1 |
NM_001318036.1 |
459 |
Silent Mutation |
GCC,GCT |
A42A |
NP_001304965.1 |
NM_001318037.1 |
459 |
Silent Mutation |
GCC,GCT |
A56A |
NP_001304966.1 |
NM_003085.4 |
459 |
Silent Mutation |
GCC,GCT |
A56A |
NP_003076.1 |
XM_006714914.3 |
459 |
Silent Mutation |
GCC,GCT |
A56A |
XP_006714977.1 |
XM_006714915.3 |
459 |
Silent Mutation |
GCC,GCT |
A56A |
XP_006714978.1 |
XM_006714916.2 |
459 |
Silent Mutation |
GCC,GCT |
A56A |
XP_006714979.1 |
XM_011534640.2 |
459 |
Silent Mutation |
GCC,GCT |
A56A |
XP_011532942.1 |
View Full Product Details