Product Details

SNP ID
rs111653954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159160514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTACCAGCCTACACTCTTTGAG[C/T]GACTTTAAAAATGGGACATAGTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF145 PubMed Links

Gene Details

Gene
RNF145
Gene Name
ring finger protein 145
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199380.1 Intron NP_001186309.1
NM_001199381.1 Intron NP_001186310.1
NM_001199382.1 Intron NP_001186311.1
NM_001199383.1 Intron NP_001186312.1
NM_144726.2 Intron NP_653327.1
XM_017009138.1 Intron XP_016864627.1

View Full Product Details