Product Details

SNP ID
rs111505970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136994957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACGATGTGCAGCTTGGGCAGGAG[A/G]CTGCAGTCGGCCAGCGTGAGCCTGT
Phenotype
MIM: 616315 MIM: 606533
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf142 PubMed Links

Gene Details

Gene
C9orf142
Gene Name
chromosome 9 open reading frame 142
There are no transcripts associated with this gene.

Gene
CLIC3
Gene Name
chloride intracellular channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004669.2 759 Silent Mutation AGC,AGT S175S NP_004660.2
XM_017015281.1 759 Silent Mutation AGC,AGT S175S XP_016870770.1
XM_017015282.1 759 Silent Mutation AGC,AGT S146S XP_016870771.1
Gene
LCNL1
Gene Name
lipocalin like 1
There are no transcripts associated with this gene.

View Full Product Details