Product Details

SNP ID
rs115488608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:33103894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACAAAGCATTGCCCAAAAGGT[A/G]AATTATGGATTTACTGCTTCCTTCT
Phenotype
MIM: 609866
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STARD13 PubMed Links

Gene Details

Gene
STARD13
Gene Name
StAR related lipid transfer domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243466.1 5167 Intron NP_001230395.1
NM_001243474.1 5167 Intron NP_001230403.1
NM_001243476.2 5167 UTR 3 NP_001230405.1
NM_052851.2 5167 UTR 3 NP_443083.1
NM_178006.3 5167 UTR 3 NP_821074.1
NM_178007.2 5167 UTR 3 NP_821075.1
XM_011535299.2 5167 UTR 3 XP_011533601.1
XM_017020834.1 5167 UTR 3 XP_016876323.1
XM_017020835.1 5167 UTR 3 XP_016876324.1

View Full Product Details