Product Details
- SNP ID
-
rs117534424
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:49310874 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATCAAGTAGTTCTTCTCGTCAGC[A/G]AACTGCTCATCATCCGTCCTTTCTT
- Phenotype
-
MIM: 612175
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAB39L
PubMed Links
Gene Details
- Gene
- CAB39L
- Gene Name
- calcium binding protein 39 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001079670.2 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
NP_001073138.1 |
NM_001287337.1 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
NP_001274266.1 |
NM_001287338.1 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
NP_001274267.1 |
NM_001287339.1 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
NP_001274268.1 |
NM_030925.3 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
NP_112187.2 |
XM_011535254.2 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
XP_011533556.1 |
XM_011535255.2 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
XP_011533557.1 |
XM_011535256.2 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
XP_011533558.1 |
XM_017020785.1 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
XP_016876274.1 |
XM_017020786.1 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
XP_016876275.1 |
XM_017020787.1 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
XP_016876276.1 |
XM_017020788.1 |
1882 |
Silent Mutation |
TTC,TTT |
F318F |
XP_016876277.1 |
View Full Product Details