Product Details

SNP ID
rs117528308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:50999063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAATTCTACCAGTGAGTGTAT[A/C]CTTCTACCCTAGCGATACAAAGAGT
Phenotype
MIM: 603695
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GUCY1B2 PubMed Links

Gene Details

Gene
GUCY1B2
Gene Name
guanylate cyclase 1 soluble subunit beta 2 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details