Product Details

SNP ID
rs117596244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51335227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGAGAGTAAATAGATAATGGC[C/T]AAAAGGGGAGAAAATATTTGCAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINE3 PubMed Links

Gene Details

Gene
SERPINE3
Gene Name
serpin family E member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101320.1 252 Intron NP_001094790.1
XM_017020707.1 252 UTR 5 XP_016876196.1

View Full Product Details