Product Details

SNP ID
rs113141374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:24685346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAACTCAGCAATAGGGAATTGGA[A/G]GAGAAATATGGCACAGACATCATTA
Phenotype
MIM: 182360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP12A PubMed Links

Gene Details

Gene
ATP12A
Gene Name
ATPase H+/K+ transporting non-gastric alpha2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185085.1 534 Silent Mutation GAA,GAG E67E NP_001172014.1
NM_001676.5 534 Silent Mutation GAA,GAG E67E NP_001667.4

View Full Product Details