Product Details

SNP ID
rs114476697
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:80038513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGCGGGGTCTCAGGTTTCCCT[C/G]ACTCCAGACTTCTCAGCCTGCCGAG
Phenotype
MIM: 611595
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
RBFA PubMed Links

Gene Details

Gene
RBFA
Gene Name
ribosome binding factor A (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171967.1 537 Silent Mutation CTC,CTG L129L NP_001165438.1
NM_024805.2 537 Silent Mutation CTC,CTG L129L NP_079081.2
Gene
TXNL4A
Gene Name
thioredoxin like 4A
There are no transcripts associated with this gene.

View Full Product Details