Product Details

SNP ID
rs117332214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116758687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTCATATATTCTTTTGGTGATT[G/T]TAAGGCTTCACATTTAATGTGAAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM162B PubMed Links

Gene Details

Gene
FAM162B
Gene Name
family with sequence similarity 162 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085480.2 Intron NP_001078949.1

View Full Product Details