Product Details

SNP ID
rs112132135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125747637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAGTTGCACCTGCAGGCATTT[C/G]AATTCAGAAAAACGTGCAGCCCTCA
Phenotype
MIM: 604674
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HEY2 PubMed Links

Gene Details

Gene
HEY2
Gene Name
hes related family bHLH transcription factor with YRPW motif 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012259.2 46 Intron NP_036391.1
XM_017010627.1 46 Intron XP_016866116.1
XM_017010628.1 46 Intron XP_016866117.1
XM_017010629.1 46 UTR 5 XP_016866118.1
Gene
LOC105377986
Gene Name
uncharacterized LOC105377986
There are no transcripts associated with this gene.

View Full Product Details