Product Details

SNP ID
rs117911374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99374405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCTAGTTCCTACCCTTATGAT[A/G]TGAGATGTTAACCTAAGGGGAAGAC
Phenotype
MIM: 605196
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ3 PubMed Links

Gene Details

Gene
COQ3
Gene Name
coenzyme Q3, methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017421.3 Intron NP_059117.3
XM_005267018.4 Intron XP_005267075.1
XM_006715500.3 Intron XP_006715563.1
XM_011535898.2 Intron XP_011534200.1
XM_011535899.1 Intron XP_011534201.1
XM_017010933.1 Intron XP_016866422.1

View Full Product Details