Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001166247.1 | 37 | Intron | NP_001159719.1 | ||
NM_021956.4 | 37 | Intron | NP_068775.1 | ||
NM_175768.3 | 37 | Intron | NP_786944.1 | ||
XM_005266945.2 | 37 | UTR 5 | XP_005267002.1 | ||
XM_005266946.3 | 37 | Intron | XP_005267003.1 | ||
XM_011535777.2 | 37 | Intron | XP_011534079.1 | ||
XM_011535778.2 | 37 | Intron | XP_011534080.1 | ||
XM_017010781.1 | 37 | Intron | XP_016866270.1 | ||
XM_017010782.1 | 37 | Intron | XP_016866271.1 |