Product Details

SNP ID
rs115184154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:166410972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTAAGACCTAAAAAGCAAATTC[A/T]TCATGGTGGAAGTCGTTCTAGCCTT
Phenotype
MIM: 601685
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RPS6KA2 PubMed Links

Gene Details

Gene
RPS6KA2
Gene Name
ribosomal protein S6 kinase A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006932.2 3984 UTR 3 NP_001006933.2
NM_001318936.1 3984 UTR 3 NP_001305865.1
NM_001318937.1 3984 UTR 3 NP_001305866.1
NM_001318938.1 3984 UTR 3 NP_001305867.1
NM_021135.5 3984 UTR 3 NP_066958.2
XM_006715549.3 3984 UTR 3 XP_006715612.1

View Full Product Details