Product Details

SNP ID
rs114946696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34753571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGTCCACATTACTGGGTGTGAC[A/G]TGGAGTCCAAGCTTTTGAGCCTGAA
Phenotype
MIM: 605680
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAZ1A PubMed Links

Gene Details

Gene
BAZ1A
Gene Name
bromodomain adjacent to zinc finger domain 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013448.2 3303 Silent Mutation CAC,CAT H1536H NP_038476.2
NM_182648.1 3303 Silent Mutation CAC,CAT H1504H NP_872589.1
XM_011536374.1 3303 Silent Mutation CAC,CAT H1443H XP_011534676.1
XM_011536376.2 3303 Silent Mutation CAC,CAT H1059H XP_011534678.2
XM_017020942.1 3303 Silent Mutation CAC,CAT H1027H XP_016876431.1

View Full Product Details