Product Details

SNP ID
rs113953690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:38255058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTCTCCCAGCTTCTCGTCGACC[C/G]TGATTGGCCATGTTCTCTGCGGCAC
Phenotype
MIM: 616845
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLEC14A PubMed Links

Gene Details

Gene
CLEC14A
Gene Name
C-type lectin domain family 14 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175060.2 1313 Missense Mutation ACG,AGG T322R NP_778230.1

View Full Product Details