Product Details

SNP ID
rs117314109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50636643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTCAGTATGTGATGAAGACAG[C/T]ACTGTAATTAATATTCTCATTTTAA
Phenotype
MIM: 606439 MIM: 607203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATL1 PubMed Links

Gene Details

Gene
ATL1
Gene Name
atlastin GTPase 1
There are no transcripts associated with this gene.

Gene
SAV1
Gene Name
salvador family WW domain containing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021818.3 Intron NP_068590.1
XM_011537057.2 Intron XP_011535359.1
XM_017021568.1 Intron XP_016877057.1

View Full Product Details