Product Details

SNP ID
rs117180958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21888330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGAGGTTCCCAGTCCTTCTTCTG[G/T]TACAGACAATATTCTGGGAAAAGCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRAV12-2 PubMed Links

Gene Details

Gene
TRAV12-2
Gene Name
T cell receptor alpha variable 12-2
There are no transcripts associated with this gene.

View Full Product Details