Product Details

SNP ID
rs116309676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55369259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGTCAGGAACAGTCTCAGCACC[A/G]CAAGGACCAGCACACTGACCCATAT
Phenotype
MIM: 613515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG14 PubMed Links

Gene Details

Gene
ATG14
Gene Name
autophagy related 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014924.4 1579 UTR 3 NP_055739.2
XM_011536563.2 1579 UTR 3 XP_011534865.1

View Full Product Details