Product Details

SNP ID
rs114074295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57202651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAACTAAGAAAACATAATGTTAA[C/T]CATGGTAAACTTTGTTCAATTCCTG
Phenotype
MIM: 604469
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC5 PubMed Links

Gene Details

Gene
EXOC5
Gene Name
exocyst complex component 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006544.3 8336 UTR 3 NP_006535.1
XM_005267272.3 8336 Intron XP_005267329.1

View Full Product Details