Product Details

SNP ID
rs111501526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:61818229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATGAGCGTTGGTATTTCACCAG[G/T]CTTGTTGCATCCATCTCCACCTCCA
Phenotype
MIM: 610950
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYT16 PubMed Links

Gene Details

Gene
SYT16
Gene Name
synaptotagmin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031914.2 Intron NP_114120.2
XM_006720272.3 Intron XP_006720335.1
XM_011537219.1 Intron XP_011535521.1
XM_011537220.1 Intron XP_011535522.1
XM_011537221.2 Intron XP_011535523.1
XM_011537222.2 Intron XP_011535524.1
XM_011537223.2 Intron XP_011535525.1
XM_011537224.2 Intron XP_011535526.1
XM_011537225.2 Intron XP_011535527.1
XM_011537227.2 Intron XP_011535529.2
XM_011537228.2 Intron XP_011535530.1
XM_011537229.2 Intron XP_011535531.1
XM_017021693.1 Intron XP_016877182.1
XM_017021694.1 Intron XP_016877183.1
XM_017021695.1 Intron XP_016877184.1
XM_017021696.1 Intron XP_016877185.1
XM_017021697.1 Intron XP_016877186.1
XM_017021698.1 Intron XP_016877187.1

View Full Product Details