Product Details

SNP ID
rs115012331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64345815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCTAGCCACTGTGGGTTTAGCA[A/G]GGATAAGCACTTGACCTAAACTGGG
Phenotype
MIM: 601663
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESR2 PubMed Links

Gene Details

Gene
ESR2
Gene Name
estrogen receptor 2
There are no transcripts associated with this gene.

Gene
TEX21P
Gene Name
testis expressed 21, pseudogene
There are no transcripts associated with this gene.

View Full Product Details