Product Details

SNP ID
rs111477155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74884684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGCCGCCATCTATTCTAGTTC[A/G]AAAACATACTATGCTTAGAATTTGC
Phenotype
MIM: 126063 MIM: 615094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLST PubMed Links

Gene Details

Gene
DLST
Gene Name
dihydrolipoamide S-succinyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244883.1 Intron NP_001231812.1
NM_001933.4 Intron NP_001924.2
Gene
PROX2
Gene Name
prospero homeobox 2
There are no transcripts associated with this gene.

View Full Product Details