Product Details
- SNP ID
-
rs117049774
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:90397433 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGCCAGGGAGCTTCATTTATTG[A/G]TTCCCCTTGGAGCTGGACTTGGTCG
- Phenotype
-
MIM: 114180
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CALM1
PubMed Links
Gene Details
- Gene
- CALM1
- Gene Name
- calmodulin 1
View Full Product Details