Product Details

SNP ID
rs117056704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113596642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTTCAGGCCAGCGGTGGAGTC[A/G]GCTAGCCACGATGACTTTTGCCCGG
Phenotype
MIM: 189902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFDP1 PubMed Links

Gene Details

Gene
TFDP1
Gene Name
transcription factor Dp-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007111.4 Intron NP_009042.1
XM_005268327.1 Intron XP_005268384.1
XM_005268328.2 Intron XP_005268385.1
XM_005268331.1 Intron XP_005268388.1
XM_011537518.2 Intron XP_011535820.1
XM_011537519.1 Intron XP_011535821.1
XM_011537520.2 Intron XP_011535822.1
XM_017020717.1 Intron XP_016876206.1
XM_017020718.1 Intron XP_016876207.1
XM_017020719.1 Intron XP_016876208.1
XM_017020720.1 Intron XP_016876209.1
XM_017020721.1 Intron XP_016876210.1
XM_017020722.1 Intron XP_016876211.1

View Full Product Details