Product Details

SNP ID
rs115520168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:145859901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCAGTGAAAACAGCTGTCCTC[A/G]GGAATATCCCCTTCCTGCAAGAGAC
Phenotype
MIM: 615762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRXCR2 PubMed Links

Gene Details

Gene
GRXCR2
Gene Name
glutaredoxin and cysteine rich domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080516.1 579 Silent Mutation CCC,CCT P193P NP_001073985.1
XM_017009708.1 579 Silent Mutation CCC,CCT P97P XP_016865197.1

View Full Product Details