Product Details
- SNP ID
-
rs114682029
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52567068 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCCTCTGTGGTCTTGGTCTGCCC[A/G]CTCTGGGTGCTGCCAGAGCTGCCTG
- Phenotype
-
MIM: 608248
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT74
PubMed Links
Gene Details
- Gene
- KRT74
- Gene Name
- keratin 74
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175053.3 |
1539 |
Silent Mutation |
|
|
NP_778223.2 |
View Full Product Details