Product Details

SNP ID
rs113795327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53100710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCCTTCTCCTATTCCTCCTCC[A/T]GTTGCAGAGGAGAATCCTAAGGAGA
Phenotype
MIM: 146735 MIM: 601311
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IGFBP6 PubMed Links

Gene Details

Gene
IGFBP6
Gene Name
insulin like growth factor binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002178.2 Intron NP_002169.1
XM_017019264.1 Intron XP_016874753.1
Gene
SOAT2
Gene Name
sterol O-acyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details