Product Details

SNP ID
rs116170001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53191553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCTTGGGTAAGTGTCCCTCCCT[C/T]CTTCAGAGAGTGGGACTGTCTGCCT
Phenotype
MIM: 147559
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGB7 PubMed Links

Gene Details

Gene
ITGB7
Gene Name
integrin subunit beta 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000889.2 2632 UTR 3 NP_000880.1
XM_005268851.3 2632 UTR 3 XP_005268908.1
XM_005268852.4 2632 UTR 3 XP_005268909.1
XM_006719376.3 2632 UTR 3 XP_006719439.1
Gene
ZNF740
Gene Name
zinc finger protein 740
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004304.3 2632 Intron NP_001004304.1
XM_006719344.2 2632 Intron XP_006719407.1
XM_011538194.2 2632 Intron XP_011536496.1
XM_017019181.1 2632 Intron XP_016874670.1

View Full Product Details