Product Details
- SNP ID
-
rs116062585
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:54371702 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGATCCTGGGGCTGGCCCCAGTCC[A/G]TTCTCCATGGAAACTGTTGTTGGGG
- Phenotype
-
MIM: 606383
MIM: 609124
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GPR84
PubMed Links
Gene Details
- Gene
- GPR84
- Gene Name
- G protein-coupled receptor 84
There are no transcripts associated with this gene.
- Gene
- LOC102724050
- Gene Name
- uncharacterized LOC102724050
There are no transcripts associated with this gene.
- Gene
- ZNF385A
- Gene Name
- zinc finger protein 385A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130967.2 |
641 |
Silent Mutation |
AAC,AAT |
N145N |
NP_001124439.1 |
NM_001130968.2 |
641 |
Intron |
|
|
NP_001124440.1 |
NM_001290001.1 |
641 |
Intron |
|
|
NP_001276930.1 |
NM_001290002.1 |
641 |
Silent Mutation |
AAC,AAT |
N125N |
NP_001276931.1 |
NM_001290004.1 |
641 |
Silent Mutation |
AAC,AAT |
N125N |
NP_001276933.1 |
NM_015481.2 |
641 |
Silent Mutation |
AAC,AAT |
N125N |
NP_056296.1 |
XM_005268783.4 |
641 |
Silent Mutation |
AAC,AAT |
N125N |
XP_005268840.1 |
XM_006719342.2 |
641 |
Silent Mutation |
AAC,AAT |
N125N |
XP_006719405.1 |
XM_011538168.2 |
641 |
Silent Mutation |
AAC,AAT |
N178N |
XP_011536470.1 |
XM_011538169.2 |
641 |
Silent Mutation |
AAC,AAT |
N141N |
XP_011536471.1 |
XM_011538170.1 |
641 |
Silent Mutation |
AAC,AAT |
N108N |
XP_011536472.1 |
XM_011538171.2 |
641 |
Intron |
|
|
XP_011536473.1 |
XM_017019174.1 |
641 |
Silent Mutation |
AAC,AAT |
N125N |
XP_016874663.1 |
View Full Product Details