Product Details

SNP ID
rs112388637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68854860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATTCATCAATATTTATCAGCA[C/T]CAATTTTTTTTTTTAAGACAGAGTC
Phenotype
MIM: 114860 MIM: 164785
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPM PubMed Links

Gene Details

Gene
CPM
Gene Name
carboxypeptidase M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005502.2 2952 UTR 3 NP_001005502.1
NM_001874.4 2952 UTR 3 NP_001865.1
NM_198320.3 2952 UTR 3 NP_938079.1
Gene
MDM2
Gene Name
MDM2 proto-oncogene
There are no transcripts associated with this gene.

View Full Product Details