Product Details

SNP ID
rs115801787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71610204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTATATGATGTTATGAGAATCTGG[C/T]AGGGCCTAGAAGCAGGCCAAACAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFC3H1 PubMed Links

Gene Details

Gene
ZFC3H1
Gene Name
zinc finger C3H1-type containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144982.4 6553 UTR 3 NP_659419.3

View Full Product Details