Product Details

SNP ID
rs111917302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71849941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCTGGAAAAGGCTAAGTTTCCC[C/T]TGAAGGGTGTTGATCCAGTATTGCA
Phenotype
MIM: 612662
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRS2P2 PubMed Links

Gene Details

Gene
MRS2P2
Gene Name
MRS2 pseudogene 2
There are no transcripts associated with this gene.

Gene
TBC1D15
Gene Name
TBC1 domain family member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146213.1 Intron NP_001139685.2
NM_001146214.1 Intron NP_001139686.1
NM_022771.4 Intron NP_073608.4
XM_006719564.2 Intron XP_006719627.1
XM_006719565.3 Intron XP_006719628.1
XM_006719566.2 Intron XP_006719629.1
XM_006719567.2 Intron XP_006719630.1
XM_011538679.1 Intron XP_011536981.1
XM_011538680.1 Intron XP_011536982.1
XM_011538681.2 Intron XP_011536983.1
XM_011538682.1 Intron XP_011536984.1
XM_017019824.1 Intron XP_016875313.1
XM_017019825.1 Intron XP_016875314.1
XM_017019826.1 Intron XP_016875315.1
XM_017019827.1 Intron XP_016875316.1
XM_017019828.1 Intron XP_016875317.1

View Full Product Details