Product Details
- SNP ID
-
rs116739226
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:76028288 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGTATACATATATACATATGAAA[C/T]AAGTTTTTAAATGTTGGCAGCATAT
- Phenotype
-
MIM: 605335
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PHLDA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146439266,rs201526762] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PHLDA1
- Gene Name
- pleckstrin homology like domain family A member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007350.3 |
3072 |
UTR 3 |
|
|
NP_031376.3 |
View Full Product Details