Product Details

SNP ID
rs111415746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76053212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTAAAACATTATTTACCTTCAGG[A/G]GGGGCAAAAAAGTTAAAGAAAGAGT
Phenotype
MIM: 164060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAP1L1 PubMed Links

Gene Details

Gene
NAP1L1
Gene Name
nucleosome assembly protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006719425.3 1308 Silent Mutation CCC,CCT P240P XP_006719488.1
XM_011538390.2 1308 Silent Mutation CCC,CCT P303P XP_011536692.2
XM_011538393.2 1308 Silent Mutation CCC,CCT P303P XP_011536695.1
XM_017019338.1 1308 Silent Mutation CCC,CCT P303P XP_016874827.1
XM_017019339.1 1308 Silent Mutation CCC,CCT P240P XP_016874828.1
XM_017019340.1 1308 Silent Mutation CCC,CCT P240P XP_016874829.1

View Full Product Details