Product Details
- SNP ID
-
rs117748424
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:79774864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTATCTTATGCAACTTTAATTAT[C/G]GTTGAGTACTATCAAGTGAAAACTG
- Phenotype
-
MIM: 602021
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP1R12A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34341780] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP1R12A
- Gene Name
- protein phosphatase 1 regulatory subunit 12A
View Full Product Details