Product Details

SNP ID
rs111635889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104457215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACGCATCTCAGCACTTCCAGA[A/C]CAACTCCGGCACCTTCCACACCCCT
Phenotype
MIM: 610128
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHST11 PubMed Links

Gene Details

Gene
CHST11
Gene Name
carbohydrate (chondroitin 4) sulfotransferase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173982.1 302 UTR 5 NP_001167453.1
NM_018413.5 302 UTR 5 NP_060883.1
XM_017019369.1 302 Intron XP_016874858.1

View Full Product Details