Product Details

SNP ID
rs115710862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196354465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGATTGACTTCACACGTTTGAG[C/T]GATTTGGGCTTCAGCTATGACTTTG
Phenotype
MIM: 616379
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBXN7 PubMed Links

Gene Details

Gene
UBXN7
Gene Name
UBX domain protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015562.1 3713 UTR 3 NP_056377.1
XM_011512670.2 3713 UTR 3 XP_011510972.1
XM_011512671.2 3713 UTR 3 XP_011510973.1

View Full Product Details