Product Details

SNP ID
rs117004505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112085489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAGGAAAGAAAAATAGTAACCA[C/T]TGGGGTCTGCAGATAGGCCTAACTC
Phenotype
MIM: 602690 MIM: 606659
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf57 PubMed Links

Gene Details

Gene
C11orf57
Gene Name
chromosome 11 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082969.1 Intron NP_001076438.1
NM_001082970.1 Intron NP_001076439.1
NM_001301017.1 Intron NP_001287946.1
NM_001301019.1 Intron NP_001287948.1
NM_001301021.1 Intron NP_001287950.1
NM_018195.3 Intron NP_060665.3
Gene
SDHD
Gene Name
succinate dehydrogenase complex subunit D
There are no transcripts associated with this gene.

Gene
TIMM8B
Gene Name
translocase of inner mitochondrial membrane 8 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012459.2 Intron NP_036591.2

View Full Product Details