Product Details

SNP ID
rs116368960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119171414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTCTTTCCCAGGATGAGGTGGG[A/G]CCACCATTTGCCCAGGGCCTCTTGG
Phenotype
MIM: 607784 MIM: 611947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCG4 PubMed Links

Gene Details

Gene
ABCG4
Gene Name
ATP binding cassette subfamily G member 4
There are no transcripts associated with this gene.

Gene
NLRX1
Gene Name
NLR family member X1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282143.1 492 Missense Mutation GAC,GGC D4G NP_001269072.1
NM_001282144.1 492 Missense Mutation GAC,GGC D4G NP_001269073.1
NM_001282358.1 492 Missense Mutation GAC,GGC D4G NP_001269287.1
NM_024618.3 492 Missense Mutation GAC,GGC D4G NP_078894.2
XM_005271669.1 492 Missense Mutation GAC,GGC D4G XP_005271726.1
XM_005271670.1 492 UTR 5 XP_005271727.1
XM_005271671.1 492 UTR 5 XP_005271728.1
XM_005271672.1 492 UTR 5 XP_005271729.1
XM_006718904.1 492 Missense Mutation GAC,GGC D4G XP_006718967.1
XM_011542980.1 492 Missense Mutation GAC,GGC D4G XP_011541282.1
XM_017018294.1 492 Missense Mutation GAC,GGC D4G XP_016873783.1

View Full Product Details