Product Details

SNP ID
rs117373613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102397843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGCTTAATAAACACAGCATTA[C/T]GCTATAGTTTTGACTCTATAGAATT
Phenotype
MIM: 606356
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM123 PubMed Links

Gene Details

Gene
TMEM123
Gene Name
transmembrane protein 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052932.2 2072 UTR 3 NP_443164.2

View Full Product Details