Product Details
- SNP ID
-
rs113272282
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:95620915 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGGCCGCCCGCGCCTGGCCGGGG[C/G]AGCGTCGCTGCCACGGGATCTTCGC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPR150
PubMed Links
Gene Details
- Gene
- GPR150
- Gene Name
- G protein-coupled receptor 150
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_199243.1 |
640 |
Missense Mutation |
CAG,GAG |
Q214E |
NP_954713.1 |
View Full Product Details