Product Details
- SNP ID
-
rs115945411
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:132483122 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTTATAAAAAGACAAATGTACA[C/T]ATTTACACACGTCCTTATGTGTTCA
- Phenotype
-
MIM: 147575
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C5orf56
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11347983] are located under a probe and SNP(s) [rs5871455] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C5orf56
- Gene Name
- chromosome 5 open reading frame 56
There are no transcripts associated with this gene.
- Gene
- IRF1
- Gene Name
- interferon regulatory factor 1
View Full Product Details