Product Details

SNP ID
rs113633692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134738833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTCAGTCTCCCGCCCATCACC[G/T]TGAATCTTCAGGGTCATTCTTCCCT
Phenotype
MIM: 601118
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CAMLG PubMed Links

Gene Details

Gene
CAMLG
Gene Name
calcium modulating ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001745.3 Intron NP_001736.1
XM_011543653.1 Intron XP_011541955.1

View Full Product Details