Product Details

SNP ID
rs112484230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139361340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAACTTATCCCAAAGGGAAAAAA[A/G]TACATATATTTCCAGTAGGAGATTT
Phenotype
MIM: 605604 MIM: 603790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAIP2 PubMed Links

Gene Details

Gene
PAIP2
Gene Name
poly(A) binding protein interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033112.2 Intron NP_001028284.1
NM_016480.4 Intron NP_057564.3
XM_011543428.1 Intron XP_011541730.1
XM_017009522.1 Intron XP_016865011.1
XM_017009523.1 Intron XP_016865012.1
Gene
SLC23A1
Gene Name
solute carrier family 23 member 1
There are no transcripts associated with this gene.

View Full Product Details