Product Details

SNP ID
rs115025368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8490225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGAGTGGTTGGTTTTCCAGGGG[A/G]TCTGCCGAGGAAGCGTGACTTCCAT
Phenotype
MIM: 160994 MIM: 606466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNRNPM PubMed Links

Gene Details

Gene
HNRNPM
Gene Name
heterogeneous nuclear ribonucleoprotein M
There are no transcripts associated with this gene.

Gene
PRAM1
Gene Name
PML-RARA regulated adaptor molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032152.4 2157 Silent Mutation GAC,GAT D659D NP_115528.4
XM_005272502.2 2157 Silent Mutation GAC,GAT D696D XP_005272559.1
XM_011528352.2 2157 Silent Mutation GAC,GAT D698D XP_011526654.1
XM_011528353.2 2157 Silent Mutation GAC,GAT D661D XP_011526655.1

View Full Product Details