Product Details

SNP ID
rs115635730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7630872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGTGATCTCTTCTTCCTGCCAG[A/G]GGGATGGAGGAGGCTGGATTCTTGT
Phenotype
MIM: 614770 MIM: 601717 MIM: 610850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCP2 PubMed Links

Gene Details

Gene
PCP2
Gene Name
Purkinje cell protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271830.1 Intron NP_001258759.1
NM_174895.2 Intron NP_777555.1
XM_006722639.2 Intron XP_006722702.1
XM_017026249.1 Intron XP_016881738.1
XM_017026250.1 Intron XP_016881739.1
XM_017026251.1 Intron XP_016881740.1
XM_017026252.1 Intron XP_016881741.1
Gene
PET100
Gene Name
PET100 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171155.1 Intron NP_001164626.1
Gene
STXBP2
Gene Name
syntaxin binding protein 2
There are no transcripts associated with this gene.

Gene
XAB2
Gene Name
XPA binding protein 2
There are no transcripts associated with this gene.

View Full Product Details